Methyl 6-chloro-4-fluoro-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

79836-500MG 79836-1G 79836-5G
77340-296EA 890.95 USD
77340-296 77358-698 77382-590
Methyl 6-chloro-4-fluoro-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥95%
Methyl 6-chloro-4-fluoro-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₅ClF₄O₂S
MW: 312.668 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768178
CAS Number: 2901105-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR