Methyl 5-fluoro-6-methoxy-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

79834-500MG 79834-1G 79834-5G
77339-344EA 579.89 USD
77339-344 77358-696 77382-588
Methyl 5-fluoro-6-methoxy-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥97%
Methyl 5-fluoro-6-methoxy-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₈F₄O₃S
MW: 308.249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768176
CAS Number: 2901099-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR