2-(Benzyloxy)-5-bromo-3-chloropyridine ≥97%

Supplier: AOBChem USA

Warning

79821-500MG 79821-1G 79821-5G 79821-25G
77334-492EA 160.68 USD
77334-492 77345-818 77370-470 77391-202
2-(Benzyloxy)-5-bromo-3-chloropyridine ≥97%
2-(Benzyloxy)-5-bromo-3-chloropyridine

Precaución: for Research Use Only

Formula: C₁₂H₉BrClNO
MW: 298.563 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29113547
CAS Number: 1289121-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR