5-Bromo-3-chloro-2-(2-methoxyethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

79820-500MG 79820-1G 79820-5G 79820-25G
77334-490EA 99.91 USD
77334-490 77342-898 77370-468 77382-586
5-Bromo-3-chloro-2-(2-methoxyethoxy)pyridine ≥97%
5-Bromo-3-chloro-2-(2-methoxyethoxy)pyridine

Precaución: for Research Use Only

Formula: C₈H₉BrClNO₂
MW: 266.52 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29113549
CAS Number: 1289268-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR