6-Chloro-3,3'-bipyridine ≥95%

Supplier: AOBChem USA

Warning

79766-500MG 79766-1G 79766-5G
77339-968EA 694.22 USD
77339-968 77358-690 77382-584
6-Chloro-3,3'-bipyridine ≥95%
6-Chloro-3,3'-bipyridine

Precaución: for Research Use Only

Formula: C₁₀H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD15475284
CAS Number: 101001-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR