2-Chloro-5-(m-tolyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

79761-500MG 79761-1G 79761-5G 79761-25G
77334-488EA 178.19 USD
77334-488 77346-702 77370-466 77391-200
2-Chloro-5-(m-tolyl)pyridine ≥95%
2-Chloro-5-(m-tolyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClN
MW: 203.667 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11849246
CAS Number: 163563-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR