2,3'-Difluoro-[1,1'-biphenyl]-4-amine ≥97%

Supplier: AOBChem USA

Warning

79753-500MG 79753-1G 79753-5G
77338-006EA 457.32 USD
77338-006 77358-686 77382-580
2,3'-Difluoro-[1,1'-biphenyl]-4-amine ≥97%
2,3'-Difluoro-[1,1'-biphenyl]-4-amine

Precaución: for Research Use Only

Formula: C₁₂H₉F₂N
MW: 205.203 g/mol
Storage Temperature: Ambient
CAS Number: 1214385-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR