2-Fluoro-4'-(trifluoromethyl)-[1,1'-biphenyl]-4-amine ≥97%

Supplier: AOBChem USA

Warning

79752-500MG 79752-1G 79752-5G
77336-766EA 393.46 USD
77336-766 77358-684 77382-578
2-Fluoro-4'-(trifluoromethyl)-[1,1'-biphenyl]-4-amine ≥97%
2-Fluoro-4'-(trifluoromethyl)-[1,1'-biphenyl]-4-amine

Precaución: for Research Use Only

Formula: C₁₃H₉F₄N
MW: 255.211 g/mol
Storage Temperature: Ambient
CAS Number: 856898-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR