2-Fluoro-2'-methyl-[1,1'-biphenyl]-4-amine ≥97%

Supplier: AOBChem USA

Warning

79751-500MG 79751-1G 79751-5G
77336-764EA 393.46 USD
77336-764 77358-682 77382-576
2-Fluoro-2'-methyl-[1,1'-biphenyl]-4-amine ≥97%
2-Fluoro-2'-methyl-[1,1'-biphenyl]-4-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₂FN
MW: 201.239 g/mol
Storage Temperature: Ambient
CAS Number: 1493031-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR