2-Fluoro-3'-methyl-[1,1'-biphenyl]-4-amine ≥97%

Supplier: AOBChem USA

Warning

79750-500MG 79750-1G 79750-5G
77335-732EA 363.59 USD
77335-732 77358-680 77382-574
2-Fluoro-3'-methyl-[1,1'-biphenyl]-4-amine ≥97%
2-Fluoro-3'-methyl-[1,1'-biphenyl]-4-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₂FN
MW: 201.239 g/mol
Storage Temperature: Ambient
CAS Number: 1208082-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR