2,4'-Difluoro-[1,1'-biphenyl]-4-amine ≥97%

Supplier: AOBChem USA

Warning

79748-500MG 79748-1G 79748-5G 79748-25G
77334-482EA 161.71 USD
77334-482 77346-008 77370-460 77391-194
2,4'-Difluoro-[1,1'-biphenyl]-4-amine ≥97%
2,4'-Difluoro-[1,1'-biphenyl]-4-amine

Precaución: for Research Use Only

Formula: C₁₂H₉F₂N
Storage Temperature: Ambient
CAS Number: 1214374-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR