5-Ethoxy-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

79711-500MG 79711-1G 79711-5G 79711-25G
77334-480EA 116.39 USD
77334-480 77343-470 77370-458 77382-570
5-Ethoxy-2-methoxybenzoic acid ≥97%
5-Ethoxy-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD30831502
CAS Number: 1368080-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR