1-(5-Chloro-2-methoxypyridin-4-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

79546-500MG 79546-1G 79546-5G 79546-25G
77334-478EA 231.75 USD
77334-478 77358-674 77370-454 77391-192
1-(5-Chloro-2-methoxypyridin-4-yl)-2,2-difluoroethanone ≥95%
1-(5-Chloro-2-methoxypyridin-4-yl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₆ClF₂NO₂
MW: 221.589 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768008
CAS Number: 2229093-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR