3-Chloro-2-methoxy-4-(trimethylsilyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

79530-500MG 79530-1G 79530-5G 79530-25G
77334-474EA 263.68 USD
77334-474 77358-670 77370-452 77391-190
3-Chloro-2-methoxy-4-(trimethylsilyl)pyridine ≥95%
3-Chloro-2-methoxy-4-(trimethylsilyl)pyridine

Precaución: for Research Use Only

Formula: C₉H₁₄ClNOSi
Storage Temperature: Ambient
MDL Number: MFCD30637303
CAS Number: 2273145-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR