3-Chloro-4-(trimethylsilyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

79526-500MG 79526-1G 79526-25G 79526-5G
77334-472EA 300.76 USD
77334-472 77358-668 77391-188 77382-562
3-Chloro-4-(trimethylsilyl)pyridine ≥95%
3-Chloro-4-(trimethylsilyl)pyridine

Precaución: for Research Use Only

Formula: C₈H₁₂ClNSi
MW: 185.726 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22042128
CAS Number: 77332-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR