2-(3-Bromo-2-chloro-6-fluoro-5-iodophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

79417-500MG 79417-1G 79417-5G 79417-25G
77334-460EA 136.99 USD
77334-460 77344-568 77370-438 77391-180
2-(3-Bromo-2-chloro-6-fluoro-5-iodophenyl)-1,3-dioxolane ≥97%
2-(3-Bromo-2-chloro-6-fluoro-5-iodophenyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₉H₆BrClFIO₂
MW: 407.403 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767971
CAS Number: 2901084-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR