2-(3-Bromo-2-chloro-6-fluoro-5-(methylthio)phenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

79243-500MG 79243-1G 79243-5G 79243-25G
77334-456EA 122.57 USD
77334-456 77344-016 77370-434 77382-544
2-(3-Bromo-2-chloro-6-fluoro-5-(methylthio)phenyl)-1,3-dioxolane ≥97%
2-(3-Bromo-2-chloro-6-fluoro-5-(methylthio)phenyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₁₀H₉BrClFO₂S
MW: 327.598 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767830
CAS Number: 2901105-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR