1-Fluoro-4-(methoxymethyl)-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

78979-1G 78979-5G 78979-25G
77342-040EA 111.24 USD
77342-040 77370-432 77382-542
1-Fluoro-4-(methoxymethyl)-2-methylbenzene ≥97%
1-Fluoro-4-(methoxymethyl)-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁FO
MW: 154.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767696
CAS Number: 1890507-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR