6-Bromo-4-methoxybenzo[d][1,3]dioxole ≥95%

Supplier: AOBChem USA

Warning

78901-500MG 78901-1G 78901-5G
77334-454EA 326.51 USD
77334-454 77358-654 77382-540
6-Bromo-4-methoxybenzo[d][1,3]dioxole ≥95%
6-Bromo-4-methoxybenzo[d][1,3]dioxole

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
MW: 231.043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22555991
CAS Number: 91511-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR