2-(4-Fluorobenzyl)-1,3,5-trimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

78793-500MG 78793-1G 78793-5G 78793-25G
77334-452EA 99.91 USD
77334-452 77342-896 77370-428 77382-536
2-(4-Fluorobenzyl)-1,3,5-trimethylbenzene ≥97%
2-(4-Fluorobenzyl)-1,3,5-trimethylbenzene

Precaución: for Research Use Only

Formula: C₁₆H₁₇F
MW: 228.3045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767659
CAS Number: 2361012-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR