2-(4-Chlorobenzyl)-1,3,5-trimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

78792-1G 78792-5G 78792-25G
77341-376EA 82.4 USD
77341-376 77370-426 77382-534
2-(4-Chlorobenzyl)-1,3,5-trimethylbenzene ≥95%
2-(4-Chlorobenzyl)-1,3,5-trimethylbenzene

Precaución: for Research Use Only

Formula: C₁₆H₁₇Cl
Storage Temperature: Ambient
MDL Number: MFCD34767658
CAS Number: 954134-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR