4-Bromo-2,6-dimethoxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

78706-500MG 78706-1G
77340-418EA 1033.09 USD
77340-418 77358-652
4-Bromo-2,6-dimethoxybenzonitrile ≥97%
4-Bromo-2,6-dimethoxybenzonitrile

Precaución: for Research Use Only

Formula: C₉H₈BrNO₂
MW: 242.069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28132246
CAS Number: 1400503-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR