5-Bromo-2-phenoxyaniline ≥97%

Supplier: AOBChem USA

Warning

78672-500MG 78672-1G 78672-5G 78672-25G
77334-448EA 143.17 USD
77334-448 77344-990 77370-422 77391-178
5-Bromo-2-phenoxyaniline ≥97%
5-Bromo-2-phenoxyaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO
MW: 264.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08687050
CAS Number: 56966-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR