1-(4-Bromophenoxy)-2-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

78666-500MG 78666-1G 78666-5G 78666-25G
77334-446EA 193.64 USD
77334-446 77358-650 77370-420 77391-176
1-(4-Bromophenoxy)-2-nitrobenzene ≥97%
1-(4-Bromophenoxy)-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₁₂H₈BrNO₃
MW: 294.101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00474965
CAS Number: 56966-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR