2-Ethoxy-3-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

78610-500MG 78610-1G 78610-5G 78610-25G
77334-436EA 260.59 USD
77334-436 77358-646 77370-410 77391-170
2-Ethoxy-3-(trifluoromethyl)benzoic acid ≥97%
2-Ethoxy-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34767576
CAS Number: 2386866-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR