3,6-Difluoro-2-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

78571-500MG 78571-1G 78571-5G 78571-25G
77334-432EA 242.05 USD
77334-432 77358-644 77370-406 77391-166
3,6-Difluoro-2-iodobenzaldehyde ≥97%
3,6-Difluoro-2-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃F₂IO
MW: 267.999 g/mol
Storage Temperature: Ambient
CAS Number: 1208077-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR