4'-Methyl-[1,1'-biphenyl]-3-amine ≥97%

Supplier: AOBChem USA

Warning

78455-500MG 78455-1G 78455-5G 78455-25G
77334-424EA 118.45 USD
77334-424 77343-576 77370-400 77382-514
4'-Methyl-[1,1'-biphenyl]-3-amine ≥97%
4'-Methyl-[1,1'-biphenyl]-3-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₃N
MW: 183.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424665
CAS Number: 400751-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR