3'-Fluoro-[1,1'-biphenyl]-3-amine ≥97%

Supplier: AOBChem USA

Warning

78452-500MG 78452-1G 78452-5G 78452-25G
77334-422EA 151.41 USD
77334-422 77345-404 77370-398 77391-160
3'-Fluoro-[1,1'-biphenyl]-3-amine ≥97%
3'-Fluoro-[1,1'-biphenyl]-3-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₀FN
MW: 187.213 g/mol
Storage Temperature: Ambient
CAS Number: 400751-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR