4'-(Trifluoromethyl)-[1,1'-biphenyl]-3-amine ≥95%

Supplier: AOBChem USA

Warning

78451-500MG 78451-1G 78451-5G 78451-25G
77334-420EA 133.9 USD
77334-420 77344-420 77370-396 77391-158
4'-(Trifluoromethyl)-[1,1'-biphenyl]-3-amine ≥95%
4'-(Trifluoromethyl)-[1,1'-biphenyl]-3-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₃N
MW: 237.22 g/mol
Storage Temperature: Ambient
CAS Number: 400747-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR