4-Methyl-2-(pyridin-4-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

78449-500MG 78449-1G 78449-5G
77334-418EA 340.93 USD
77334-418 77358-634 77382-512
4-Methyl-2-(pyridin-4-yl)benzaldehyde ≥97%
4-Methyl-2-(pyridin-4-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD24238609
CAS Number: 1554525-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR