4',5-Dimethyl-[1,1'-biphenyl]-2-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

78440-500MG 78440-1G 78440-5G 78440-25G
77334-416EA 202.91 USD
77334-416 77358-632 77370-394 77391-156
4',5-Dimethyl-[1,1'-biphenyl]-2-carbaldehyde ≥97%
4',5-Dimethyl-[1,1'-biphenyl]-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₄O
MW: 210.271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24238603
CAS Number: 216442-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR