5-Methyl-[1,1'-biphenyl]-2-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

78439-500MG 78439-1G 78439-5G 78439-25G
77334-414EA 126.69 USD
77334-414 77344-194 77370-392 77382-510
5-Methyl-[1,1'-biphenyl]-2-carbaldehyde ≥95%
5-Methyl-[1,1'-biphenyl]-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₂O
MW: 196.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24232548
CAS Number: 934691-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR