2'-Chloro-5'-methyl-[1,1'-biphenyl]-3-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

78432-500MG 78432-1G 78432-25G 78432-5G
77334-410EA 301.79 USD
77334-410 77358-628 77391-154 77382-506
2'-Chloro-5'-methyl-[1,1'-biphenyl]-3-carbaldehyde ≥95%
2'-Chloro-5'-methyl-[1,1'-biphenyl]-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO
MW: 230.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767516
CAS Number: 1999897-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR