(2-Chloro-6-fluoro-5-iodo-3-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

78299-500MG 78299-1G 78299-5G 78299-25G
77334-404EA 149.35 USD
77334-404 77345-324 77370-386 77391-150
(2-Chloro-6-fluoro-5-iodo-3-methylphenyl)(methyl)sulfane ≥97%
(2-Chloro-6-fluoro-5-iodo-3-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₇ClFIS
MW: 316.562 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767472
CAS Number: 2901084-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR