(4-Chloro-2-fluoro-5-methyl-1,3-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

78298-500MG 78298-1G 78298-25G 78298-5G
77334-402EA 292.52 USD
77334-402 77358-624 77391-148 77382-502
(4-Chloro-2-fluoro-5-methyl-1,3-phenylene)bis(methylsulfane) ≥97%
(4-Chloro-2-fluoro-5-methyl-1,3-phenylene)bis(methylsulfane)

Precaución: for Research Use Only

Formula: C₉H₁₀ClFS₂
MW: 236.757 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767471
CAS Number: 2901105-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR