4-Chloro-2-fluoro-5-methyl-3-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

78296-500MG 78296-1G 78296-5G 78296-25G
77334-400EA 224.54 USD
77334-400 77358-622 77370-384 77391-146
4-Chloro-2-fluoro-5-methyl-3-(methylthio)benzaldehyde ≥97%
4-Chloro-2-fluoro-5-methyl-3-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈ClFOS
Storage Temperature: Ambient
MDL Number: MFCD34767469
CAS Number: 2901109-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR