4-Chloro-2-fluoro-5-methyl-3-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

78294-500MG 78294-1G 78294-5G 78294-25G
77334-398EA 131.84 USD
77334-398 77344-378 77370-382 77391-144
4-Chloro-2-fluoro-5-methyl-3-(methylthio)benzoic acid ≥97%
4-Chloro-2-fluoro-5-methyl-3-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂S
Storage Temperature: Ambient
MDL Number: MFCD34767467
CAS Number: 2901084-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR