2-Chloro-4-fluoro-5-iodo-1,3-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

78291-500MG 78291-1G 78291-5G
77337-604EA 426.42 USD
77337-604 77358-618 77382-500
2-Chloro-4-fluoro-5-iodo-1,3-dimethylbenzene ≥97%
2-Chloro-4-fluoro-5-iodo-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₇ClFI
Storage Temperature: Ambient
MDL Number: MFCD34767464
CAS Number: 2901105-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR