4-Fluoro-N,N-dimethylbenzothioamide ≥97%

Supplier: AOBChem USA

Warning

78178-500MG 78178-1G 78178-5G 78178-25G
77334-388EA 156.56 USD
77334-388 77345-630 77370-374 77391-138
4-Fluoro-N,N-dimethylbenzothioamide ≥97%
4-Fluoro-N,N-dimethylbenzothioamide

Precaución: for Research Use Only

Formula: C₉H₁₀FNS
MW: 183.2458 g/mol
Storage Temperature: Ambient
CAS Number: 1664368-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR