diethyl 2-(3,5-dimethoxybenzylidene)malonate ≥95%

Supplier: AOBChem USA

Warning

78120-500MG 78120-1G 78120-5G 78120-25G
77334-386EA 202.91 USD
77334-386 77358-606 77370-372 77391-136
diethyl 2-(3,5-dimethoxybenzylidene)malonate ≥95%
diethyl 2-(3,5-dimethoxybenzylidene)malonate

Precaución: for Research Use Only

Formula: C₁₆H₂₀O₆
MW: 308.3264 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767341
CAS Number: 5859-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR