2,6-Diethoxy-3-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

78102-500MG 78102-1G 78102-5G 78102-25G
77334-382EA 150.38 USD
77334-382 77345-366 77370-368 77391-132
2,6-Diethoxy-3-iodopyridine ≥97%
2,6-Diethoxy-3-iodopyridine

Precaución: for Research Use Only

Formula: C₉H₁₂INO₂
Storage Temperature: Ambient
MDL Number: MFCD34767336
CAS Number: 2901081-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR