6-Chloro-2-hydroxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

78065-500MG 78065-1G 78065-25G 78065-5G
77334-376EA 301.79 USD
77334-376 77358-600 77391-128 77382-484
6-Chloro-2-hydroxy-3-methylbenzoic acid ≥97%
6-Chloro-2-hydroxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MW: 186.5923 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21234538
CAS Number: 872276-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR