6-Chloro-2-hydroxy-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

78064-500MG 78064-1G 78064-5G
77334-374EA 310.03 USD
77334-374 77358-598 77382-482
6-Chloro-2-hydroxy-3-methylbenzaldehyde ≥97%
6-Chloro-2-hydroxy-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
MW: 170.5929 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21281097
CAS Number: 1310800-53-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR