4-(3-Fluorophenoxy)-2-methyl-1-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

77962-500MG 77962-1G 77962-5G 77962-25G
77334-372EA 99.91 USD
77334-372 77342-892 77370-360 77382-478
4-(3-Fluorophenoxy)-2-methyl-1-nitrobenzene ≥97%
4-(3-Fluorophenoxy)-2-methyl-1-nitrobenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₀FNO₃
Storage Temperature: Ambient
MDL Number: MFCD34767305
CAS Number: 2432903-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR