4-(4-Bromophenoxy)-2-methyl-1-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

77960-500MG 77960-1G 77960-5G 77960-25G
77334-370EA 121.54 USD
77334-370 77343-882 77370-358 77382-476
4-(4-Bromophenoxy)-2-methyl-1-nitrobenzene ≥97%
4-(4-Bromophenoxy)-2-methyl-1-nitrobenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrNO₃
MW: 308.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12069426
CAS Number: 163258-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR