4-Bromo-5-(methylthio)thiophene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

77860-500MG 77860-1G 77860-5G 77860-25G
77334-368EA 252.35 USD
77334-368 77358-594 77370-356 77391-126
4-Bromo-5-(methylthio)thiophene-2-carboxylic acid ≥97%
4-Bromo-5-(methylthio)thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅BrO₂S₂
MW: 253.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18839113
CAS Number: 172461-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR