2-Iodo-1,3-dimethoxy-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

77841-500MG 77841-1G 77841-5G
77335-654EA 361.53 USD
77335-654 77358-592 77382-472
2-Iodo-1,3-dimethoxy-5-methylbenzene ≥95%
2-Iodo-1,3-dimethoxy-5-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁IO₂
MW: 278.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28793671
CAS Number: 54812-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR