1-(Benzo[b]thiophen-2-yl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

77825-500MG 77825-1G 77825-5G 77825-25G
77334-364EA 264.71 USD
77334-364 77358-588 77370-354 77391-122
1-(Benzo[b]thiophen-2-yl)-2,2,2-trifluoroethanone ≥97%
1-(Benzo[b]thiophen-2-yl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₅F₃OS
Storage Temperature: Ambient
MDL Number: MFCD07380761
CAS Number: 75277-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR