4-Bromo-5-ethoxy-2-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

77743-500MG 77743-1G 77743-5G 77743-25G
77334-362EA 144.2 USD
77334-362 77345-026 77370-352 77391-120
4-Bromo-5-ethoxy-2-fluorophenol ≥97%
4-Bromo-5-ethoxy-2-fluorophenol

Precaución: for Research Use Only

Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28738551
CAS Number: 1807139-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR