(2-Chloro-5-fluoro-3-isobutoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

77728-500MG 77728-1G 77728-5G
77339-246EA 569.59 USD
77339-246 77358-586 77382-468
(2-Chloro-5-fluoro-3-isobutoxyphenyl)(methyl)sulfane ≥97%
(2-Chloro-5-fluoro-3-isobutoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClFOS
Storage Temperature: Ambient
MDL Number: MFCD34767222
CAS Number: 2901084-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR